3-Formyl-2-methyl-5-nitroindole 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Methyl-5-nitro-1H-indole-3-carboxaldehyde

OR6332-1G OR6332-5G OR6332-500MG
APOSOR6332-1GEA 79.9 EUR
APOSOR6332-1G APOSOR6332-5G APOSOR6332-500MG
3-Formyl-2-methyl-5-nitroindole 95+%
3-Formyl-2-methyl-5-nitroindole
Formula: C₁₀H₈N₂O₃
MW: 204,19 g/mol
Melting Pt: 305…307 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00216481
CAS Number: 3558-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR