4-Oxocyclohexanecarboxylic acid 95%
Supplier: Apollo Scientific
|
Danger
|
OR6345-1G
OR6345-25G
OR6345-5G
APOSOR6345-1GEA
29
EUR
APOSOR6345-1G
APOSOR6345-25G
APOSOR6345-5G
4-Oxocyclohexanecarboxylic acid 95%
4-Oxocyclohexanecarboxylic acid
Formula:
C₇H₁₀O₃ MW: 142,16 g/mol Melting Pt: 67…71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00102035 CAS Number: 874-61-3 EINECS: 677-204-7 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...