5H-Imidazo[1,2-b]pyrazole-2-carboxylic acid 95%
Supplier: Apollo Scientific
|
Warning
|
OR6352-100MG
OR6352-250MG
APOSOR6352-100MGEA
217
EUR
APOSOR6352-100MG
APOSOR6352-250MG
5H-Imidazo[1,2-b]pyrazole-2-carboxylic acid 95%
5H-Imidazo[1,2-b]pyrazole-2-carboxylic acid
Formula:
C₆H₅N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD08056315 CAS Number: 914637-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...