Boc-L-norvaline 98%
Supplier: Apollo Scientific
Synonyms:
Boc-Nva-OH, N-(tert-Butoxycarbonyl)-L-norvaline, (2S)-2-Aminopentanoic acid, N-BOC protected, N-Boc-L-norvaline
OR6368-1G
OR6368-5G
APOSOR6368-1GEA
49.5
EUR
APOSOR6368-1G
APOSOR6368-5G
Boc-L-norvaline 98%
Boc-L-norvaline
Formula:
C₁₀H₁₉NO₄ MW: 217,265 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037268 CAS Number: 53308-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...