5-Methyl-1,3-oxazole-4-carbonyl chloride

Supplier: Apollo Scientific
Danger

OR6378-1G OR6378-250MG OR6378-500MG
APOSOR6378-1GEA 519 EUR
APOSOR6378-1G APOSOR6378-250MG APOSOR6378-500MG
5-Methyl-1,3-oxazole-4-carbonyl chloride
5-Methyl-1,3-oxazole-4-carbonyl chloride
Formula: C₅H₄ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD08056322
CAS Number: 914637-76-6
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR