Methyl-5-bromo-6-hydroxynicotinate 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 5-bromo-6-hydroxynicotinate
OR6398-5G
APOSOR6398-5GEA
79.9
EUR
APOSOR6398-5G
Methyl-5-bromo-6-hydroxynicotinate 95%
Methyl 5-bromo-6-hydroxynicotinate
Formula:
C₇H₆BrNO₃ MW: 232,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08235247 CAS Number: 381247-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...