3-(Pyrimidin-2-yl)bromobenzene 95%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(3-Bromophenyl)pyrimidine

OR6404-1G OR6404-250MG
APOSOR6404-1GEA 364 EUR
APOSOR6404-1G APOSOR6404-250MG
3-(Pyrimidin-2-yl)bromobenzene 95%
3-(Pyrimidin-2-yl)bromobenzene
Formula: C₁₀H₇BrN₂
MW: 235,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08056330
CAS Number: 885468-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR