2-Bromo-3-hydroxypyridine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Bromo-3-pyridinol

OR6410-100G OR6410-5G OR6410-10G
APOSOR6410-100GEA 53.8 EUR
APOSOR6410-100G APOSOR6410-5G APOSOR6410-10G
2-Bromo-3-hydroxypyridine 98%
2-Bromo-3-hydroxypyridine
Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 182…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006220
CAS Number: 6602-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR