3-Chloro-4-morpholino-1,2,5-thiadiazole 97%

Supplier: Apollo Scientific

Synonyms: Timolol Related Compound F, 4-(4-Chloro-1,2,5-thiadiazol-3-yl)morpholine

OR6417-25G OR6417-5G
APOSOR6417-25GEA 78.6 EUR
APOSOR6417-25G APOSOR6417-5G
3-Chloro-4-morpholino-1,2,5-thiadiazole 97%
3-Chloro-4-morpholino-1,2,5-thiadiazole
Formula: C₆H₈ClN₃OS
MW: 205,66 g/mol
Melting Pt: 44…47 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00274160
CAS Number: 30165-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR