3-Bromo-6,7,8,9-tetrahydro-5H-imidazo[1,2-a]azepine

Supplier: Apollo Scientific
Warning

OR6419-1G OR6419-250MG
APOSOR6419-1GEA 430 EUR
APOSOR6419-1G APOSOR6419-250MG
3-Bromo-6,7,8,9-tetrahydro-5H-imidazo[1,2-a]azepine
3-Bromo-6,7,8,9-tetrahydro-5H-imidazo[1,2-a]azepine
Formula: C₈H₁₁BrN₂
Storage Temperature: Ambient
MDL Number: MFCD08235254
CAS Number: 701298-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR