Propargyl chloride 97%
Supplier: Apollo Scientific
OR6425-25G
OR6425-100G
APOSOR6425-25GEA
109
EUR
APOSOR6425-25G
APOSOR6425-100G
Propargyl chloride 97%
Propagyl chloride
Formula:
C₃H₃Cl Boiling Pt: 58 °C Density: 1,03 g/cm³ (25 °C) Flash Pt: 19 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000980 CAS Number: 624-65-7 UN: 3286 ADR: 3+6.1+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...