2-Bromo-6-methylpyridine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Bromo-2-picoline, 2-Bromo-6-picoline
OR6451-100G
OR6451-25G
APOSOR6451-100GEA
53.8
EUR
APOSOR6451-100G
APOSOR6451-25G
2-Bromo-6-methylpyridine 98%
2-Bromo-6-methylpyridine
Formula:
C₆H₆BrN Boiling Pt: 102…103 °C/20mmHg Density: 1,512 g/cm³ (25 °C) Flash Pt: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040743 CAS Number: 5315-25-3 EINECS: 226-173-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...