6-Chloro-1-hexanol 97+%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Chlorohexan-1-ol, 1-Chloro-6-hydroxyhexane, 6-Chlorohexanol

OR6510-100G OR6510-250G OR6510-50G
APOSOR6510-100GEA 50.9 EUR
APOSOR6510-100G APOSOR6510-250G APOSOR6510-50G
6-Chloro-1-hexanol 97+%
6-Chloro-1-hexanol
Formula: C₆H₁₃ClO
Boiling Pt: 108…112 °C/14mmHg
Density: 1,024 g/cm³ (25 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002984
CAS Number: 2009-83-8
EINECS: 217-925-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR