1,2,3,4-Tetrahydroisoquinoline 97%
Supplier: Apollo Scientific
|
Warning
|
OR6521-100G
OR6521-25G
OR6521-500G
OR6521-250MG
APOSOR6521-100GEA
116
EUR
APOSOR6521-100G
APOSOR6521-25G
APOSOR6521-500G
APOSOR6521-250MG
1,2,3,4-Tetrahydroisoquinoline 97%
1,2,3,4-Tetrahydroisoquinoline
Formula:
C₉H₁₁N MW: 133,19 g/mol Boiling Pt: 232…233 °C Melting Pt: –30 °C Density: 1,049 g/cm³ (25 °C) Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006896 CAS Number: 91-21-4 EINECS: 202-050-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...