N-Phthaloylglycine 98%
Supplier: Apollo Scientific
|
Warning
|
OR6526-100G
OR6526-10G
OR6526-500G
APOSOR6526-100GEA
81.2
EUR
APOSOR6526-100G
APOSOR6526-10G
APOSOR6526-500G
N-Phthaloylglycine 98%
N-Phthaloylglycine
Formula:
C₁₀H₇NO₄ Melting Pt: 193…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005900 CAS Number: 4702-13-0 EINECS: 225-177-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...