Fluorene-9-carboxylic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: 9H-Fluorene-9-carboxylic acid

OR6528-100G OR6528-10G OR6528-25G
APOSOR6528-100GEA 132 EUR
APOSOR6528-100G APOSOR6528-10G APOSOR6528-25G
Fluorene-9-carboxylic acid 95%
Fluorene-9-carboxylic acid
Formula: C₁₄H₁₀O₂
MW: 210,23 g/mol
Melting Pt: 226…232 °C
Flash Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00001136
CAS Number: 1989-33-9
EINECS: 217-866-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR