Diethyl (2-phenylethyl)malonate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Diethyl 2-phenethylmalonate
OR6546-25G
OR6546-5G
OR6546-100G
APOSOR6546-25GEA
270
EUR
APOSOR6546-25G
APOSOR6546-5G
APOSOR6546-100G
Diethyl (2-phenylethyl)malonate 98%
Diethyl (2-phenylethyl)malonate
Formula:
C₁₅H₂₀O₄ MW: 264,32 g/mol Boiling Pt: 112…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026887 CAS Number: 6628-68-8 EINECS: 229-610-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...