Benzo[b]thiophene 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Thianaphthene, 1-Benzothiophene
OR6572-5G
OR6572-100G
OR6572-25G
APOSOR6572-5GEA
21.9
EUR
APOSOR6572-5G
APOSOR6572-100G
APOSOR6572-25G
Benzo[b]thiophene 97%
Benzo[b]thiophene
Formula:
C₈H₆S MW: 134,2 g/mol Boiling Pt: 221…222 °C Melting Pt: 28…33 °C Density: 1,149 g/cm³ (25 °C) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005864 CAS Number: 95-15-8 EINECS: 202-395-7 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...