3-(2-Furyl)acrylic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(Furan-2-yl)acrylic acid

OR6587-25G OR6587-100G OR6587-1G OR6587-5G
APOSOR6587-25GEA 51 EUR
APOSOR6587-25G APOSOR6587-100G APOSOR6587-1G APOSOR6587-5G
3-(2-Furyl)acrylic acid 98%
3-(2-Furyl)acrylic acid
Formula: C₇H₆O₃
MW: 138,12 g/mol
Boiling Pt: 285…287 °C
Melting Pt: 139…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00003257
CAS Number: 539-47-9
EINECS: 208-718-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR