Bromoacetamide 96%

Supplier: Apollo Scientific
Danger

OR6592-5G OR6592-25G
APOSOR6592-5GEA 26.1 EUR
APOSOR6592-5G APOSOR6592-25G
Bromoacetamide 96%
Bromoacetamide
Formula: BrCH₂CONH₂
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00008025
CAS Number: 683-57-8
UN: 2923
ADR: 8+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR