3'-Acetamidoacetophenone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Acetamidoacetophenone
OR6598-25G
OR6598-5G
OR6598-100G
APOSOR6598-25GEA
24.8
EUR
APOSOR6598-25G
APOSOR6598-5G
APOSOR6598-100G
3'-Acetamidoacetophenone 98%
3'-Acetamidoacetophenone
Formula:
C₁₀H₁₁NO₂ MW: 177.20 g/mol Melting Pt: 125…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00032278 CAS Number: 7463-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...