3-Methoxythiophene

Supplier: Apollo Scientific
Warning

OR6628-100G OR6628-25G OR6628-5G
APOSOR6628-100GEA 109 EUR
APOSOR6628-100G APOSOR6628-25G APOSOR6628-5G
3-Methoxythiophene
3-Methoxythiophene
Formula: C₅H₆OS
Boiling Pt: 80…82 °C/65mmHg
Density: 1,143 g/cm³ (25 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00043894
CAS Number: 17573-92-1
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR