4-Chlorophenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Chlorobenzeneboronic acid, Dihydroxy(4-chlorophenyl)borane, (4-Chlorophenyl)boronic acid, (4-Chlorophenyl)metaboric acid

OR6643-100G OR6643-25G OR6643-500G
APOSOR6643-100GEA 84.1 EUR
APOSOR6643-100G APOSOR6643-25G APOSOR6643-500G
4-Chlorophenylboronic acid 98%
4-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
MW: 156,38 g/mol
Melting Pt: 284…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR