2,3-Dihydro-1-(methylsulphonyl)-(1H)-indole-5-amine 95+%
Supplier: Apollo Scientific
|
Warning
|
OR6776-1G
OR6776-250MG
APOSOR6776-1GEA
94.5
EUR
APOSOR6776-1G
APOSOR6776-250MG
2,3-Dihydro-1-(methylsulphonyl)-(1H)-indole-5-amine 95+%
2,3-Dihydro-1-(methylsulphonyl)-(1H)-indole-5-amine
Formula:
C₉H₁₂N₂O₂S Melting Pt: 115…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD01319362 CAS Number: 299921-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...