Triethyl-2-phosphonopropionate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Diethyl [1-(ethoxycarbonyl)ethyl]phosphonate, Ethyl 2-(diethoxyphosphoryl)propanoate
OR6820-25G
OR6820-5G
APOSOR6820-25GEA
75.7
EUR
APOSOR6820-25G
APOSOR6820-5G
Triethyl-2-phosphonopropionate 97%
Triethyl 2-phosphonopropionate
Formula:
C₉H₁₉O₅P MW: 238,22 g/mol Boiling Pt: 143…144 °C/12mmHg Density: 1,11 g/cm³ (25 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009159 CAS Number: 3699-66-9 EINECS: 223-033-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...