1-(2-Cyanophenyl)piperazine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Piperazin-1-yl)benzonitrile, 1-(2-Cyanophenyl)piperazine, 2-(1-Piperazinyl)benzonitrile
OR6867-100G
OR6867-25G
OR6867-5G
APOSOR6867-100GEA
941
EUR
APOSOR6867-100G
APOSOR6867-25G
APOSOR6867-5G
1-(2-Cyanophenyl)piperazine 98%
1-(2-Cyanophenyl)piperazine
Formula:
C₁₁H₁₃N₃ MW: 187,25 g/mol Boiling Pt: 314…315 °C Density: 1,115 g/cm³ (25 °C) Flash Pt: 47,8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040793 CAS Number: 111373-03-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...