1-Heptylpiperazine 97%
Supplier: Apollo Scientific
|
Warning
|
OR6894-10G
OR6894-2G
OR6894-5G
APOSOR6894-10GEA
284
EUR
APOSOR6894-10G
APOSOR6894-2G
APOSOR6894-5G
1-Heptylpiperazine 97%
1-Heptylpiperazine
Formula:
C₁₁H₂₄N₂ Boiling Pt: 106…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075184 CAS Number: 82502-77-0 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...