1-(2-Thien-2-ylethyl)piperazine 97%

Supplier: Apollo Scientific
Warning

OR6920-1G OR6920-5G
APOSOR6920-1GEA 69.6 EUR
APOSOR6920-1G APOSOR6920-5G
1-(2-Thien-2-ylethyl)piperazine 97%
1-(2-Thien-2-ylethyl)piperazine
Formula: C₁₀H₁₆N₂S
Boiling Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD01320903
CAS Number: 461046-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR