1-Benzyl-4-hydroxypiperidine 98%

Supplier: Apollo Scientific
Danger

Synonyms: 1-Benzylpiperidin-4-ol

OR6935-100G OR6935-500G
APOSOR6935-100GEA 147 EUR
APOSOR6935-100G APOSOR6935-500G
1-Benzyl-4-hydroxypiperidine 98%
1-Benzyl-4-hydroxypiperidine
Formula: C₁₂H₁₇NO
MW: 191,27 g/mol
Boiling Pt: 127…128 °C/2mmHg
Melting Pt: 61…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00006503
CAS Number: 4727-72-4
EINECS: 225-226-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR