2-(3-Hydroxycarbonyl-6-pyridyl)malondialdehyde 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 6-(1,3-Dioxoprop-2-yl)nicotinic acid

OR6985-1G OR6985-5G
APOSOR6985-1GEA 371 EUR
APOSOR6985-1G APOSOR6985-5G
2-(3-Hydroxycarbonyl-6-pyridyl)malondialdehyde 95+%
2-(3-Hydroxycarbonyl-6-pyridyl)malondialdehyde
Formula: C₉H₇NO₄
MW: 193,16 g/mol
Melting Pt: 275 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00216532
CAS Number: 212755-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR