Thymoquinone
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
p-Mentha-3,6-diene-2,5-dione, 2-Isopropyl-5-methylcyclohexa-2,5-diene-1,4-dione, 5-Isopropyl-2-methyl-1,4-benzoquinone
OR70000-1G
APOSOR70000-1GEA
29.2
EUR
APOSOR70000-1G
Thymoquinone
Thymoquinone
Formula:
C₁₀H₁₂O₂ MW: 164,20 g/mol Boiling Pt: 230…232 °C Melting Pt: 45…47 °C Flash Pt: 104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001602 CAS Number: 490-91-5 EINECS: 207-721-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...