2-Iodobiphenyl 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Iodo-1,1'-biphenyl
OR70057-100G
OR70057-25G
OR70057-5G
APOSOR70057-100GEA
366
EUR
APOSOR70057-100G
APOSOR70057-25G
APOSOR70057-5G
2-Iodobiphenyl 95%
2-Iodobiphenyl
Formula:
C₁₂H₉I MW: 280,11 g/mol Boiling Pt: 300 °C Density: 1,59 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039396 CAS Number: 2113-51-1 EINECS: 218-303-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...