2-Pentynoic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Pent-2-ynoic acid
OR70066-1G
OR70066-25G
OR70066-5G
APOSOR70066-1GEA
80
EUR
APOSOR70066-1G
APOSOR70066-25G
APOSOR70066-5G
2-Pentynoic acid 97%
2-Pentynoic acid
Formula:
C₅H₆O₂ MW: 98,10 g/mol Melting Pt: 47…53 °C Flash Pt: 96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014370 CAS Number: 5963-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...