3-Nitrobenzoylacetonitrile

Supplier: Apollo Scientific
Warning

OR7010-1G OR7010-5G
APOSOR7010-1GEA 58 EUR
APOSOR7010-1G APOSOR7010-5G
3-Nitrobenzoylacetonitrile
3-Nitrobenzoylacetonitrile
Formula: C₉H₆N₂O₃
Storage Temperature: Ambient
CAS Number: 21667-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR