4-Chloro-3-hydroxyphenylboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: (4-Chloro-3-hydroxyphenyl)boronic acid

OR70139-1G OR70139-250MG
APOSOR70139-1GEA 126 EUR
APOSOR70139-1G APOSOR70139-250MG
4-Chloro-3-hydroxyphenylboronic acid
4-Chloro-3-hydroxyphenylboronic acid
Formula: C₆H₆BClO₃
MW: 172,3740 g/mol
Melting Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD10697413
CAS Number: 915201-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR