2-Amino-5-chloro-3-iodobenzonitrile

Supplier: Apollo Scientific
Danger

OR70215-1G OR70215-5G
APOSOR70215-1GEA 128 EUR
APOSOR70215-1G APOSOR70215-5G
2-Amino-5-chloro-3-iodobenzonitrile
2-Amino-5-chloro-3-iodobenzonitrile
Formula: C₇H₄ClIN₂
Storage Temperature: Ambient
MDL Number: MFCD09800802
CAS Number: 1000577-48-9
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR