4-Aminobenzamide

Supplier: Apollo Scientific
Warning

OR7078-100G OR7078-500G
APOSOR7078-100GEA 40.6 EUR
APOSOR7078-100G APOSOR7078-500G
4-Aminobenzamide
4-Aminobenzamide
Formula: C₇H₈N₂O
Melting Pt: 181…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00007999
CAS Number: 2835-68-9
EINECS: 220-612-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR