Methyl-5-(methylsulphonyl)indole-2-carboxylate 95+%

Supplier: Apollo Scientific

Synonyms: Methyl 5-(methylsulphonyl)-1H-indole-2-carboxylate

OR7112-1G OR7112-5G
APOSOR7112-1GEA 74.1 EUR
APOSOR7112-1G APOSOR7112-5G
Methyl-5-(methylsulphonyl)indole-2-carboxylate 95+%
Methyl-5-(methylsulphonyl)indole-2-carboxylate
Formula: C₁₁H₁₁NO₄S
MW: 253,28 g/mol
Melting Pt: 176…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00216478
CAS Number: 205873-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR