3-(tert-Butoxymethyl)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

OR7174-1G OR7174-5G
APOSOR7174-1GEA 29 EUR
APOSOR7174-1G APOSOR7174-5G
3-(tert-Butoxymethyl)phenylboronic acid 98%
3-(tert-Butoxymethyl)phenylboronic acid
Formula: C₁₁H₁₇BO₃
MW: 208,07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03788411
CAS Number: 858364-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR