4-(tert-Butoxymethyl)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: (4-(tert-Butoxymethyl)phenyl)boronic acid

OR7176-1G OR7176-250MG OR7176-5G
APOSOR7176-1GEA 24.8 EUR
APOSOR7176-1G APOSOR7176-250MG APOSOR7176-5G
4-(tert-Butoxymethyl)phenylboronic acid 98%
4-(tert-Butoxymethyl)phenylboronic acid
Formula: C₁₁H₁₇BO₃
MW: 208,07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03701685
CAS Number: 1024017-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR