4-Nonylphenylboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 4-N-Nonylphenylboronic acid, 4-n-Nonylbenzeneboronic acid, FAAH/MAGL-IN-4

OR7244-1G OR7244-5G
APOSOR7244-1GEA 158 EUR
APOSOR7244-1G APOSOR7244-5G
4-Nonylphenylboronic acid
4-Nonylphenylboronic acid
Formula: C₁₅H₂₅BO₂
Melting Pt: 82…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093070
CAS Number: 256383-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR