2-Propoxyphenylboronic acid 96%
Supplier: Apollo Scientific
|
Warning
|
OR7255-1G
OR7255-5G
APOSOR7255-1GEA
94.4
EUR
APOSOR7255-1G
APOSOR7255-5G
2-Propoxyphenylboronic acid 96%
2-Propoxyphenylboronic acid
Formula:
C₉H₁₃BO₃ MW: 180,0087 g/mol Melting Pt: 65…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427191 CAS Number: 134896-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...