N-Cyclohexylmaleimide 98%
Supplier: Apollo Scientific
|
Danger
|
OR7279-100G
OR7279-25G
APOSOR7279-100GEA
408
EUR
APOSOR7279-100G
APOSOR7279-25G
N-Cyclohexylmaleimide 98%
N-Cyclohexylmaleimide
Formula:
C₁₀H₁₃NO₂ MW: 179,22 g/mol Melting Pt: 81…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043904 CAS Number: 1631-25-0 UN: 2811 ADR: 6.1,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...