4-(4-Chlorophenyl)aniline
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4'-Chloro-[1,1'-biphenyl]-4-amine, 4-Amino-4'-chlorobiphenyl
OR7343-1G
OR7343-250MG
APOSOR7343-1GEA
272
EUR
APOSOR7343-1G
APOSOR7343-250MG
4-(4-Chlorophenyl)aniline
4-(4-Chlorophenyl)aniline
Formula:
C₁₂H₁₀ClN MW: 203,6700 g/mol Melting Pt: 127…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00477989 CAS Number: 135-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...