3'-Chlorobiphenyl-4-carboxylic acid 98%
Supplier: Apollo Scientific
|
Warning
|
OR7363-1G
OR7363-5G
OR7363-10G
APOSOR7363-1GEA
161
EUR
APOSOR7363-1G
APOSOR7363-5G
APOSOR7363-10G
3'-Chlorobiphenyl-4-carboxylic acid 98%
3'-Chlorobiphenyl-4-carboxylic acid
Formula:
C₁₃H₉ClO₂ MW: 232,67 g/mol Melting Pt: 249…250 °C Storage Temperature: Ambient |
CAS Number:
5728-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...