1-Naphthoyl chloride 98%

Supplier: Apollo Scientific
Danger

OR7500-100G OR7500-250G
APOSOR7500-100GEA 172 EUR
APOSOR7500-100G APOSOR7500-250G
1-Naphthoyl chloride 98%
1-Naphthoyl chloride
Formula: C₁₁H₇ClO
MW: 190,63 g/mol
Boiling Pt: 190 °C/35mmHg
Melting Pt: 16…19 °C
Density: 1,265 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004002
CAS Number: 879-18-5
EINECS: 212-903-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR