3-Butyne-2-one 98%

Supplier: Apollo Scientific
Danger

Synonyms: But-3-yn-2-one

OR7548-10G OR7548-100G
APOSOR7548-10GEA 168 EUR
APOSOR7548-10G APOSOR7548-100G
3-Butyne-2-one 98%
3-Butyne-2-one
Formula: C₄H₄O
MW: 68,08 g/mol
Boiling Pt: 84…86 °C
Density: 0,87 g/cm³ (25 °C)
Flash Pt: –1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008775
CAS Number: 1423-60-5
EINECS: 215-834-2
UN: 1992
ADR: 3+6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR