3-Amino-5-tert-butyl-1H-pyrazole 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Amino-5-tert-butylpyrazole

OR7580-1G OR7580-5G
APOSOR7580-1GEA 0 EUR
APOSOR7580-1G APOSOR7580-5G
3-Amino-5-tert-butyl-1H-pyrazole 98%
3-Amino-5-tert-butyl-1H-pyrazole
Formula: C₇H₁₃N₃
MW: 139,20 g/mol
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00067987
CAS Number: 82560-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR