4-Bromostyrene 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Bromo-4-vinylbenzene
OR7587-25G
OR7587-5G
OR7587-100G
APOSOR7587-25GEA
48
EUR
APOSOR7587-25G
APOSOR7587-5G
APOSOR7587-100G
4-Bromostyrene 95%
4-Bromostyrene
Formula:
C₈H₇Br MW: 183,05 g/mol Boiling Pt: 88…89 °C/16mmHg Melting Pt: 44350 °C Density: 1,4 g/cm³ (25 °C) Flash Pt: 75 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000110 CAS Number: 2039-82-9 EINECS: 218-022-6 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...