4-Cyanopiperidine 96%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Piperidine-4-carbonitrile
OR7602-1G
OR7602-5G
APOSOR7602-1GEA
21.9
EUR
APOSOR7602-1G
APOSOR7602-5G
4-Cyanopiperidine 96%
4-Cyanopiperidine
Formula:
C₆H₁₀N₂ MW: 110,16 g/mol Boiling Pt: 121…123 °C/30mmHg Density: 0,987 g/cm³ (25 °C) Flash Pt: 105 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05022468 CAS Number: 4395-98-6 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...